Check Order Status
Agarose (Intermediate Melting/PCR Grade), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Agarose (Intermediate Melting/PCR Grade), Fisher BioReagents
For separation of low MW DNA ≤1000 bp by PCR analysis
Have Questions?
Catalog Number
Quantity
FBRBP2410-100
100 g
Catalog number
FBRBP2410-100
Price (MYR)
2,539.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,539.00
EA
Add to cart
Specifications
Chemical Name or Material
Agarose
Name Note
Intermediate Melting
Physical Form
Powder/Solid
CAS
9012-36-6
ChemAlert Storage Symbol
Gray
View more