Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™
Fisher Chemical
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1758-500500 mLPoly Bottle
Catalog number FBRBP1758-500
Price (MYR)
618.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
618.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris Hydrochloride
Name Note1M Solution, pH 8.0
CAS77-86-1
ChemAlert Storage SymbolGray
ColorUndesignated
View more
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 8.0 provide convenient and highly purified solutions.