Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Fisher Chemical
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Have Questions?
Change viewbuttonViewtableView
Quantity:
400 mL
100 mL
Packaging:
Coated Amber Glass/Poison Pack
Catalog number FBRBP1750I-400
Price (MYR)
1,123.00
EA
Add to cart
Quantity:
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Request bulk or custom format
Price (MYR)
1,123.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenol, Saturated
CAS108-95-2
ChemAlert Storage SymbolWhite
Health Hazard 2DANGER!
CAS6381-92-6
View more
Phenol, Saturated (pH 6.6/7.9, Liq.)
Phenol saturated with Tris buffer is used in DNA extraction procedures.