Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Fisher Chemical
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Poly Bottle
Catalog number FBRBP170-500
Price (MYR)
819.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
819.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcrylamide
Physical FormSolid
Acrylic Acid0.002% max.
CAS79-06-1
ChemAlert Storage SymbolBlue
View more
This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.