Check Order Status
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number
FBRBP168-500
Price (MYR)
424.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
424.00
EA
Add to cart
Specifications
Chemical Name or Material
Boric Acid
pH
3.8 to 4.8
Color
White
CAS
10043-35-3
ChemAlert Storage Symbol
Gray
View more
This high-purity electrophoresis-grade reagent is used in the preparation of Tris/Borate/EDTA electrophoresis buffers.