For solubilization and permeabilization of membranes
Have Questions?
Change view
Quantity:
500 mL
100 mL
Packaging:
View all
HDPE Bottle
Catalog number FBRBP151-500
Price (MYR)
502.00
EA
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
502.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriton™ X-100
Shelf Life2 years
ColorUndesignated
CAS9002-93-1
ChemAlert Storage SymbolGray
View more
This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.