Microbiology Media Additives: Tryptone
Microbiology Media Additives: Tryptone
Actual product may vary
Microbiology Media Additives: Tryptone
Additive for preparing bacterial nutrient media
Have Questions?
Catalog NumberQuantity
FBRBP1421-500500 g
Catalog number FBRBP1421-500
Price (MYR)
-
Quantity:
500 g
Note: This is a product of animal origin (pancreas).
Specifications
DescriptionBacterial Culture
FormDehydrated Powder
GradeMolecular genetics
Media TypeTryptone
Product TypeMicrobiology Media
For Use With (Application)Pancreatic digest of casein is used as a nitrogen source for bacteria
Packaging TypePoly Bottle
Quantity500 g
Unit SizeEA