Check Order Status
Acrylamide: Bis-Acrylamide 19:1 (Powder/Electrophoresis), Fisher BioReagents
Element Search
Structure Search
Acrylamide: Bis-Acrylamide 19:1 (Powder/Electrophoresis), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP1364-100
100 g
Catalog number
FBRBP1364-100
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
Acrylamide: Bis-Acrylamide 19:1
Physical Form
Solid
CAS
79-06-1
ChemAlert Storage Symbol
Blue
CAS
110-26-9
View more
This dry powder mix is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.