Check Order Status
Pre-Filled Bead Mill Tubes
Actual product may vary
Pre-Filled Bead Mill Tubes
For use with Bead Mill 4 and Bead Mill 24 homogenizers. May also be used with most other commercial Bead Mills.
Have Questions?
Catalog Number
Capacity (Metric)
Particle Size
Material
Product Type
FBO15565799
2 mL
2.8 mm
Ceramic
Hard Tissue Homogenizing Mix Tubes
Catalog number
FBO15565799
Price (MYR)
691.00
Special offer
691.00
EA
Add to cart
Capacity (Metric):
2 mL
Particle Size:
2.8 mm
Material:
Ceramic
Product Type:
Hard Tissue Homogenizing Mix Tubes
Price (MYR)
691.00
Special offer
691.00
EA
Add to cart
Polypropylene tubes closed with screw caps
Non-Sterile
Specifications
Product Type
Hard Tissue Homogenizing Mix Tubes
Capacity (Metric)
2 mL
Particle Size
2.8 mm
Volume Range (Metric)
300 μL to 1.8 mL
Diameter (Metric)
10.8 mm
Length (Metric)
46 mm (without cap); 49 mm (with cap)
Material
Ceramic
Dnase Free
Yes
For Use With (Application)
Used for processing brain, liver, muscle, skin, leaves
For Use With (Equipment)
Bead Mill 4 and 24 Homogenizers
RNase Free
Yes