112xx Series Advanced Ultrasonic Cleaner
Actual product may vary
112xx Series Advanced Ultrasonic Cleaner
Fisherbrand 112xx Series Advanced Ultrasonic Cleaners are more powerful than conventional cleaners. They feature a wide range of adjustable parameters for lab applications including cleaning, mixing, and degassing. Dual frequency capability allows safe quiet cleaning and mixing of sensitive samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelCapacity (Metric)Voltage
FBEFB11211FB-1121128 L115 V
FBEFB11201FB-112012.75 L115 V
FBEFB11207FB-1120712.75 L115 V
Catalog number FBEFB11211
Price (MYR)
-
Model:
FB-11211
Capacity (Metric):
28 L
Voltage:
115 V
Fisherbrand 112xx Series Advanced Ultrasonic Cleaners are more powerful than conventional cleaners. They feature a wide range of adjustable parameters for lab applications including cleaning, mixing, and degassing. Dual frequency capability allows safe quiet cleaning and mixing of sensitive samples.
  • Dual frequency: 37/80kHz
  • Use high frequency (80kHz) for longer cleaning of sensitive items
  • Low volume even at high frequency
  • Simple-to-use interface and easy-to-read display
  • Rear drain with a convenient side valve for ease-of-use
  • Use the Power Control for power regulation of special cleaning and laboratory applications
  • Pause option allows you to stop the cleaning
  • Auto Start functionality to start cleaning when a programmed temperature is reached
  • Safety feature shuts down the machine after 12 hours of continuous operation
Choose the exact cleaning mode for your needs
  • Normal: ideal for sample preparation
  • Pulse: activatable additional power for increased peak performance
  • Sweep: uniform distribution of power throughout the tank
  • Degas: efficient degassing of samples and HPLC solvents
Specifications
Voltage115 V
Capacity (English)7.3 gal.
Capacity (Metric)28 L
Dimensions (D x W x H)13.4 x 22.4 x 12.6 in.
Dimensions (D x W x H) Interior11.8 x 19.9 x 7.9 in.
For Use With (Application)Cleaning, Mixing and Degassing in Lab and Industry
ModelFB-11211
Wattage1580 w
Operating Frequency37/80 kHz
Unit SizeEach