Elmasonic Easy 180 H with Cover
Elmasonic Easy 180 H with Cover
Actual product may vary
Elmasonic Easy 180 H with Cover
The Fisherbrand Easy family include a range of 9 different device sizes with tank volumes ranging from 0.8 to 28 liters. Microprocessor-controlled ultrasonic water bath with sweep technology offers a state-of-the-art solution.
Have Questions?
Catalog number FBE16443899
Price (MYR)
-
  • Efficient 37 kHz ultrasonic high-performance transducers
  • Cleaning tank made of cavitation-resistant stainless steel
  • User-friendly and clearly arranged operating panel; sealed against liquid intrusion to protect the electronics
  • LED display for ultrasonic function
  • Rotary switch for easy preselection of the cleaning time
  • Continuous operating or short-term operation between 1 and 30 minutes
  • Permanent sweep function for optimized sound field distribution through frequency modulation
  • Dry-run protected heating
  • Temperature selectable by rotary switch; from 30°C to 80°C in increments of 5°C (for H units)
  • LED display for heating operation (for H units)
  • Detachable power cord with IEC plug
  • Selectable temperature threshold with alarm
  • Automatic safety shutdown after 8 hours of continuous operation
  • Automatic safety shutdown at a bath temperature of 90°C
  • Pulse function
Specifications
MaterialStainless Steel
DisplayLED
Frequency37 kHz
Capacity (Metric)18 L
IncludesCover
Unit SizeEach