Check Order Status
Device Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Device Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Provides a secure closure for ultrasonic cleaning baths.
Have Questions?
Catalog Number
For Use With (Equipment)
FBE1076810
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Catalog number
FBE1076810
Price (MYR)
-
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Flat design with handle
Available in a range of sizes for all X-tra ST models
Manufactured from stainless steel
Accessory item
Specifications
Type
Lid
For Use With (Equipment)
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
For Use With (Application)
Closure for X-tra ST Ultrasonic Cleaners
Includes
Handle
Shape
Flat