Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Actual product may vary
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Mini-Centrifuge 100-240V, 50/60Hz Universal Plug, Grey
Tool-free rotors and quick-spin options make the Fisherbrand Mini-Centrifuge fast and easy to use. The locking lid is functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction.
Have Questions?
Catalog NumberCapacity
FBA12-006-9016 x 1.5/2 mL
Catalog number FBA12-006-901
Price (MYR)
795.00
Special offer
1,046.00
Save 251.00 (24%)
EA
Add to cart
Capacity:
6 x 1.5/2 mL
Price (MYR)
795.00
Special offer
1,046.00
Save 251.00 (24%)
EA
Add to cart
The Fisherbrand Mini-Centrifuge is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 mL) for customized use. The centrifuge housing is made from ABS plastic and the lid is a polycarbonate plastic enabling the unit to be easily cleaned. A braking function quickly stops the unit when powered off or when the lid is opened.
  • 5 Year Warranty
  • A locking lid for added protection when the unit is in use
  • A larger lid to accommodate longer and skirted tubes, as well as taller tube lids
  • A braking function to quickly stop the unit when powered off or when the lid is opened
  • Meets the latest applicable North American & EU safety requirements
  • Weighted rubberized base eliminates potential movement and further dampens vibration and noise
  • Capacity: 6 x 1.5/2 mL Tubes (Rotor 1: Adapters for 0.5 mL and 0.2 mL Tubes), 2 places for 8 x 0.2 mL Strips or 16 Single Tubes (Rotor 2)
Specifications
Capacity6 x 1.5/2 mL
Max. Speed6,000 rpm
Max. RCF2,000 x g
RefrigeratedNo
Width (English) Exterior5.03 in.
Width (Metric) Exterior128 mm
TypeMini-Centrifuge
IncludesTwo rotors, tube rack, storage case, O-rings, tube adapters and low voltage, double insulated power adapter with four interchangeable plugs
Electrical Requirements100/240 V 50/60 Hz
Rotor IncludedYes
Dimensions (HWD)104 x 128 x 153 mm
Frequency50/60 Hz
Voltage100/240 V
Unit SizeEA