Thermolyne™ Atmosphere Controlled Ashing Furnaces
Thermolyne™ Atmosphere Controlled Ashing Furnaces
Actual product may vary
Thermolyne™ Atmosphere Controlled Ashing Furnaces
Achieve consistent temperature control with this furnace, which reaches 975°C with the standard manifold and 1093°C with the optional inconel manifold.
Have Questions?
Catalog NumberCapacity (Metric)Temperature ControlVoltageCertifications/Compliance
F6028C-6014 LDigital single setpoint w/ramp and dwell (B1)208 VCSA certified
Catalog number F6028C-60
Price (MYR)
33,110.00
EA
Add to cart
Capacity (Metric):
14 L
Temperature Control:
Digital single setpoint w/ramp and dwell (B1)
Voltage:
208 V
Certifications/Compliance:
CSA certified
Price (MYR)
33,110.00
EA
Add to cart
Achieve consistent temperature control with the Thermo Scientific™ Thermolyne™ Atmosphere Controlled Ashing Furnace, which reaches 975°C with the standard stainless-steel manifold and 1093°C with the optional inconel manifold.
Key Features:
  • Adjustable gas flowmeter/valve (0-80L/min.) on front for easy access when adjusting the airflow rate
  • Stainless-steel manifold at rear chamber prewarms incoming gases, provides a maximum temperature gradient of only ±3°C at 750°C
  • Chamber rear has a 0.95cm (0.38in.) port for monitoring chamber temperatures with independent measuring devices
  • With 0.64cm (0.25in.) I.D. or 0.96cm (0.375in.) O.D. hose barb (in chamber rear) for inert gas line
Type F6000:
  • With two dual-purpose stainless-steel trays and handle to accommodate 24 (30mL) porcelain crucibles or 38 (10mL) quartz crucibles
Type F6000-60:
  • Meets ASTM™ D3174 specifications: 3 to 4 air exchanges per min.
  • Heating rate of 8°C/min. to 500°C, 6°C/min. from 500° to 750°C
  • Holds at 750°C for two hours, then turns off
Requires:
Electrical connection requires fixed wiring (power cord not provided).
Compatible with:
  • Stainless-steel shelf (SH408X1)
  • Stainless-steel flexible exhaust tubing kit with mounting hardware (AY408X1A)
Specifications
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA certified
DescriptionF6028C-60
Capacity (Metric)14 L
Temperature (Metric)975°C
TypeSingle Setpoint
Power Consumption4000 w
Temperature ControlDigital single setpoint w/ramp and dwell (B1)
Dimensions (L x W x H) Exterior20 x 19.1 x 21 in. (51 x 49 x 53 cm)
Length (English)20 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)49 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Voltage208 V
Amperage19.2 A
Unit SizeEA