Search Thermo Fisher Scientific
| Catalog Number | Type | Description |
|---|---|---|
| F-300X also known as 00101-18195 | PEEK Wing Nut w/PEEK Ferrule | 1/16 in. |
| F-120X also known as IDEX = F-120X | One-Piece PEEK Fingertight Fitting | 1/16 in., 0.37 in. Head |
| F-130X | One-Piece PEEK Long Fingertight Fitting | 1/16 in., 0.37 in. Head |
| HYPU-467BLK | Short Head Port Plug, Black | 1/16 in., 10-32 Coned, Delrin, Black |
| U-467BLK | Short Head Port Plug, Black | 1/16 in., 10-32 Coned, Delrin, Black |
Description