SN 38
SN 38
SN 38
SN 38
DNA topoisomerase I inhibitor; antitumor
Have Questions?
Catalog NumberQuantity
EYE26845050 mg
Catalog number EYE268450
Price (MYR)
-
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialSN 38
CAS86639-52-3
Recommended StorageStore at +4°C
Quantity50 mg
Molecular FormulaC22H20N2O5
View more
Active metabolite of CPT-11 (Cat. No. 2688) that inhibits DNA topoisomerase I (IC50 values are 0.74 and 1.9 μM in P388 and Ehrlich cells respectively). Inhibits DNA and RNA synthesis (IC50 values are 0.077 and 1.3 μM respectively) but does not affect protein synthesis. Displays potent antitumor activity against a range of human tumor cell lines (IC50 values are 3.3, 13, 19 and 22 nM for HCT-116, BEL-7402, HL60 and HELA cells respectively).