PSB 069
PSB 069
PSB 069
PSB 069
Non-selective NTPDase inhibitor
Have Questions?
Catalog NumberQuantity
EYE25731010 mg
Catalog number EYE257310
Price (MYR)
8,555.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
8,555.00
EA
Add to cart
Specifications
Chemical Name or MaterialPSB 069
CAS78510-31-3
Recommended StorageStore at Room Temperature
Quantity10 mg
Molecular FormulaC20H12ClN2NaO5S
View more
Non-selective nucleoside triphosphate diphosphohydrolase (NTPDase) inhibitor. Reported to inhibit rat NTPDases 1, 2 and 3 with similar potencies.