Check Order Status
NSC 625987
Actual product may vary
NSC 625987
Cyclin-dependent kinase 4 (cdk4) inhibitor
Have Questions?
Catalog Number
Quantity
EYE215250
50 mg
Catalog number
EYE215250
Price (MYR)
343.00
EA
Add to cart
Quantity:
50 mg
Price (MYR)
343.00
EA
Add to cart
Cyclin-dependent kinase (cdk) 4 inhibitor (IC50 = 0.2 μM at cdk4/cyclin D1). Displays > 500-fold selectivity over cdk2 (IC50 > 100 μM for cdc2/cyclin A, cdk2/cyclin A and cdk2/cyclin E).
For Research Use Only
Specifications
Purity
>98%
Quantity
50 mg
Product Type
NSC 625987
Molecular Weight (g/mol)
271.33
Inhibitors
NSC 625987