PPT
PPT
PPT
PPT
Subtype-selective ERα agonist
Have Questions?
Catalog NumberQuantity
EYE14261010 mg
Catalog number EYE142610
Price (MYR)
909.00
PK
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
909.00
PK
Add to cart
Specifications
Chemical Name or MaterialPPT
CAS263717-53-9
Recommended StorageStore at +4°C
Quantity10 mg
Molecular FormulaC24H22N2O3
View more
Potent, subtype-selective estrogen receptor agonist (EC50 ∽ 200 pM); displays 410-fold selectivity for ERα over ERβ. Prevents ovariectomy-induced weight gain and loss of bone mineral density, and induces gene expression in the hypothalamus following systemic administration in vivo.