Check Order Status
Carolina™ Crayfish
Actual product may vary
Carolina™ Crayfish
Crayfish, also called crawfish or crawdads, are freshwater crustaceans resembling small lobsters.
Have Questions?
Catalog Number
Product Type
Quantity
Size
Length (English)
EYE142500
Crayfish
3/Container
Large
3.5 in.
Catalog number
EYE142500
Price (MYR)
669.00
EA
Add to cart
Product Type:
Crayfish
Quantity:
3/Container
Size:
Large
Length (English):
3.5 in.
Price (MYR)
669.00
EA
Add to cart
Not sexed
Large (3 1/2 inches or larger) and medium (about 3 inches) sizes are good for laboratory study and dissection
Small size (less than 3 inches) is ideal for the aquarium
Please order 10 days in advance
Specifications
For Use With (Application)
Good for laboratory study and dissection
Storage Requirements
18°C to 25°C
Quantity
3/Container
Size
Large
Product Type
Crayfish
Length (English)
3.5 in.