Check Order Status
Light 1163 Shoes
Actual product may vary
Light 1163 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Have Questions?
Catalog Number
Size
EYE116340
40
Catalog number
EYE116340
Price (MYR)
803.00
EA
Add to cart
Size:
40
Price (MYR)
803.00
EA
Add to cart
CE EN ISO 20347:2012 OB A, E, FO SRA
Smooth leather
White/light blue
Slip resistance PU outsole sole
Replaceable acc Wave insole (article no. 3558)
Sanitized treated
Antistatic
Heel strap non-foldable, adjustable
Breathable lining with Silver ions
Instep area with lacing
Reflective material
Specifications
Certifications/Compliance
CE EN ISO 20347:2012 OB A, E, FO SRA
Color
White, Light Blue
Material
Leather
Size
40
Sole Features
Slip-resistant
Sole Material
Polyurethane
Style
1163