Polypropylene Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY
Polypropylene Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY
Actual product may vary
Polypropylene Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
Have Questions?
Catalog NumberDimensions (L x W)
EYE11616014 x 10 in.
Catalog number EYE116160
Price (MYR)
482.00
EA
Add to cart
Dimensions (L x W):
14 x 10 in.
Price (MYR)
482.00
EA
Add to cart
  • High density polypropylene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
Specifications
FinishNot Laminated
MaterialPolypropylene
Quantity1 Ea.
Adhesive TypeNon-adhesive
DescriptionHigh density polypropylene
Mounting TypeCorner Holes
Height (English)10 in.
Max. Temperature (English)185°F
Min. Temperature (English)-40°F
Thickness0.0551 in.
Width (English)14 in.
ShapeRectangular
Temperature Range (English)-40°F to 185°F
Dimensions (L x W)14 x 10 in.
Displayed TextFLAMMABLE KEEP FLAMES AND HEAT AWAY
ColorRed on White
Unit SizeEA