Search Thermo Fisher Scientific
Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.
| Catalog Number | Length (Metric) | Diameter (Metric) | Particle Size | Packing Material |
|---|---|---|---|---|
| ES902 | 250 mm | 75 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
| ES901 | 150 mm | 50 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.
EASY-Spray ̶ Fully integrated HPLC column/temperature/emitter design
Easy Spray Design Benefits