Check Order Status
Ehrlich Aldehyde Reagent, Alcoholic, for Indole Test, Ricca Chemical
Element Search
Structure Search
Ehrlich Aldehyde Reagent, Alcoholic, for Indole Test, Ricca Chemical
For Indole Test
Have Questions?
Catalog Number
Quantity
ERW27904
120 mL
Catalog number
ERW27904
Price (MYR)
37,313.00
EA
Add to cart
Quantity:
120 mL
Request bulk or custom format
Price (MYR)
37,313.00
EA
Add to cart
Chemical Identifiers
CAS
64-17-5, 7647-01-0, 7732-18-5, 100-10-7, 7732-18-5
Specifications
Chemical Name or Material
Ehrlich Aldehyde Reagent
Color
Yellow
CAS
64-17-5
CAS Min %
70.0
CAS Max %
73.0
View more