Standard Ferrules
Standard Ferrules
Actual product may vary
Standard Ferrules
Made of high-purity materials that will not contribute background impurities for sensitive and selective detectors; designed to seat securely in a variety of chromatography tube and pipe fittings
Have Questions?
Catalog NumberMaterialSize (English) FittingDiameter (English) Inside
ERW2707715% Graphite, 85% Vespel1/8 in.1/8 in.
Catalog number ERW27077
Price (MYR)
1,474.00
EA
Add to cart
Material:
15% Graphite, 85% Vespel
Size (English) Fitting:
1/8 in.
Diameter (English) Inside:
1/8 in.
Price (MYR)
1,474.00
EA
Add to cart
For 1/16, 1/8, and 1/4 in. fittings.
100% Graphite ferrules
  • High-density and contain no binders that can off-gas or absorb analytes
  • With a smoother surface and cleaner edges than conventional graphite ferrules
  • Ferrules are very soft and can be easily deformed for an effective seal with fused silica or glass columns
  • Stable to 450°C (842°F)
Vespel™/Graphite ferrules
  • 60%/40% blend, offering the best combination of sealing performance and ease of workability
  • Seal with minimal torque and are preferred for vacuum and high-pressure uses such as GC/MS or other oxygen-sensitive detectors
  • Hard material does not deform easily and may require retightening after several temperature cycles
  • Reusable
  • Stable to 400°C (752°F)
Specifications
ColorBlack
Diameter (English)1/8 in.
Diameter (English) Inside1/8 in.
Diameter (Metric)3.175 mm
For Use With (Application)1/16 in., 1/8 in., and 1/4 in. Fittings
Size (English) Fitting1/8 in.
Material15% Graphite, 85% Vespel
TypeFerrule
Quantity10/Pk.
Unit SizeEA