Check Order Status
Inserts for 1545 mm Vials
Actual product may vary
Inserts for 1545 mm Vials
Allow for maximum sample recovery and easier sample removal.
Have Questions?
Catalog Number
Type
ERW27076
Conical Insert
Catalog number
ERW27076
Price (MYR)
1,474.00
EA
Add to cart
Type:
Conical Insert
Price (MYR)
1,474.00
EA
Add to cart
When used in conjunction with autosampler vials, these allow for maximum sample recovery and easier sample removal.
Vial inserts (0.75 mL, glass, insert type, shell style, insert 8 x 43 mm) allow the use of smaller sample volumes with standard autosampler vials.
Inserts should be chosen based on the inner diameter of the vial neck.
Specifications
Diameter (Metric) Outer
6 mm
Material
Glass
Volume (Metric)
0.25 mL
No. per Pack
1000
Height (Metric)
39 mm
Type
Conical Insert
For Use With (Equipment)
4 mL Screw Cap Vial, 15 x 45 mm