Air-Operated Stainless Steel Sample Pump
Air-Operated Stainless Steel Sample Pump
Actual product may vary
Air-Operated Stainless Steel Sample Pump
Stainless steel air-operated sample pump.
Have Questions?
Catalog NumberDescription
ERW18007BEN1800-7
Catalog number ERW18007
Price (MYR)
519.00
EA
Add to cart
Description:
BEN1800-7
Price (MYR)
519.00
EA
Add to cart
Stainless steel air-operated sample pump.
  • For 2 in. (50 mm) wells and larger.
  • Model 1800 lifts up to 1,000 feet.
  • Equipped with a 1 in. diameter pump piston for maximum flow rates at low pumping lifts.
  • Intermediate lifts require a 7/8 in. diameter pump piston that reduces the flow rate.
  • High lift applications require a 3/4 in. diameter pump piston with further reductions in pump flow rate.
Specifications
MaterialStainless Steel
TypeSample Pump
DescriptionBEN1800-7
Unit SizeEA