HM-200 Series Flask/Funnel Heating Mantle with Controller
HM-200 Series Flask/Funnel Heating Mantle with Controller
Actual product may vary
HM-200 Series Flask/Funnel Heating Mantle with Controller
The HM-200 Series Flask/Funnel Heating Mantle has an opening in its base that accommodates a wide variety of flasks and 60° funnels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Wattage
EMX5000SCE
also known as EMX5000/SCE
2000 to 5000 mL600 W
EMX1000SCE
also known as EMX1000/SCE
500 to 1000 mL240 W
Catalog number EMX5000SCE
also known as EMX5000/SCE
Price (MYR)
-
Capacity (Metric):
2000 to 5000 mL
Wattage:
600 W
Accommodates 60°C funnels, pear-shaped or round bottom flasks. Electrothermal™ EMX Series Spill-proof Electromantle is designed to accept a large range of flasks and funnel sizes for added flexibility. Stainless steel liner over the heating element providea additional electrical and mechanical protection against spills and ensures easier cleaning.
  • Ceramic fiber insulation keeps case cool.
  • Built-in power controller offers convenience.
  • Chemical-resistant polypropylene housing.
  • Funnel-shaped cavity with a base opening allows uses with 60° funnels and a wide variety of round bottom and pear shaped flasks.
  • Heating mantles have maximum element temperature of 842°F(450°C).
  • Ceramic fiber insulation keeps the polypropylene housing cool.
  • Built-in power controller with dial markings from 1 to 10 offer repeatability.
  • The ''power on'' and ''heater on'' lights indicate if the unit is on.
  • The 36101-series heating mantles with stainless steel (SS) liner over the heating element offer protection against spills and make cleaning easier.
  • The 36104-series heating mantles include a safety earth (ground) screen to provide operator safety.
  • All mantles (except 5000 mL models) feature a single support rod clamp that accepts a 1/2 in. rod.
Specifications
Capacity2000 to 5000 mL (67.6 to 169.1 oz.)
Capacity (Metric)2000 to 5000 mL
For Use WithFlasks
Dimensions (L x W x H)400 x 350 x 190 mm
Voltage230 V
Liner MaterialStainless steel screen
Power Supply DetailsUK and EU plug
Wattage600 W
Weight (Metric) Shipping5.96 kg
Product TypeHeating Mantle
Temperature (Metric)450°C
Capacity (English)67.6 to 169.1 oz.
Frequency50/60 Hz