HM-200 Series Heating Mantle
HM-200 Series Heating Mantle
Actual product may vary
HM-200 Series Heating Mantle
The HM-200 Series Heating Mantle with thermal insulating cartridge provides maximum heat transfer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
ELEEM5000CE
also known as ELEEM5000/CE
5000 mL
ELEEM1000CE
also known as ELEEM1000/CE
1000 mL
ELEEM3000CE
also known as ELEEM3000/CE
3000 mL
Catalog number ELEEM5000CE
also known as ELEEM5000/CE
Price (MYR)
13,146.00
EA
Add to cart
Capacity (Metric):
5000 mL
Price (MYR)
13,146.00
EA
Add to cart
Includes a built-in energy regulator with indicator lamps for power and heater operation. Electrothermal™ EM Series Controlled Electromantle incorporates a grounded earth screen and double fuses for added safety. Larger capacity products have multiple heating elements, enabling a more even and directed transfer of heat.
  • Chemical resistant polypropylene housing is ''safe to the touch.''
  • Quickly and safely heat round-bottom flasks to 450°C (842°F).
  • These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability.
  • An amber-colored ''heater on'' light and a clear-colored ''power on'' light indicate when the heating mantle is in use.
  • The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
  • Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use.
  • A safety earth (ground) screen and double fuses provide added safety.
Specifications
Closure TypePolypropylene Outer Case
Capacity5000 mL (169.1 oz.)
Capacity (Metric)5000 mL
Dimensions (L x W x H)400 x 350 x 190 mm
Height (English)7.5 in.
Height (Metric)190 mm
Length (Metric)400 mm
Voltage230 V
MaterialPolypropylene
Power Supply DetailsUK and EU plug
Wattage800 W
Weight (Metric) Shipping2.58 kg
Width (English)9.8 in.
Width (Metric)250 mm
Temperature (Metric)450°C
Capacity (English)169.1 oz.
Temperature (English)842°F