Check Order Status
CMU Series Uncontrolled Mantle
Actual product may vary
CMU Series Uncontrolled Mantle
Double-fused and grounded for safety. Electrothermal™ CMU Series Uncontrolled Mantle requires a controller such as MC5 or MC242.
Have Questions?
Catalog Number
Capacity (Metric)
Dimensions (L x W x H)
Wattage
Weight (Metric) Shipping
ELECMU5000E
also known as ELECMU5000/E
5000 mL
320 x 320 x 220mm
800W
2.75kg
Catalog number
ELECMU5000E
also known as ELECMU5000/E
Price (MYR)
-
Capacity (Metric):
5000 mL
Dimensions (L x W x H):
320 x 320 x 220mm
Wattage:
800W
Weight (Metric) Shipping:
2.75kg
Valuable for fume hood or remote working
Must be used with a controller; recommended controllers are MC5 and MC242
Round bottom flask capacities of between 50mL to 5000mL
“Cool-to-touch” outer casing
Element temperatures up to 450°C
Indicator lamps for power and heater operation
Replaceable insulated heater cartridge
Rugged, easy to clean powder coated aluminum casing
Non-skid feet and support rod bracket (Support rods not included)
Flexible coiled heating elements absorb shock, minimising the risk of flask breakage
Heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support
Safety features include a grounded earth stainless steel screen covering the heating element and double fuses
Specifications
Capacity (Metric)
5000 mL
Dimensions (L x W x H)
320 x 320 x 220mm
Voltage
230 V
Material
Powder-coated aluminum
Wattage
800W
Weight (Metric) Shipping
2.75kg
Temperature (Metric)
450°C
Frequency
50/60 Hz