Check Order Status
Fold Up Wall Mount Table with Hand Crank
Actual product may vary
Fold Up Wall Mount Table with Hand Crank
Mopec™ Fold Up Wall Mount Table provides a compact work area for prep, surgery, and exams.
Have Questions?
Catalog number
ELECD500
Price (MYR)
-
18 gauge stainless steel with marine edge for fluid control
Provides a compact work area
Release handle unlocks the table from an upright position to a 90 degree horizontal position
Specifications
Product Type
Fold-up Wall Mount Table
Length (English)
45 in.
Width (English)
20 in.
Length (Metric)
114.3 cm
Weight (English)
Holds up to 300 lb.
Weight (Metric)
Holds up to 136.08 Kg
Width (Metric) Sink
50.8 cm