7000 Series Ultra Precision Wire Cutters with Medium Heads
7000 Series Ultra Precision Wire Cutters with Medium Heads
Actual product may vary
7000 Series Ultra Precision Wire Cutters with Medium Heads
7000 Series Ultra Precision Wire Cutters with Medium Heads
7000 Series Ultra Precision Wire Cutters with Medium Heads
For cutting leads and soft wire
Have Questions?
Catalog NumberCapacity (English)Cut TypeHandle TypeHead Style
ECT7140E0.008 to 0.042 in.Semi-flushErgonomic longMedium Round
Catalog number ECT7140E
Price (MYR)
724.00
EA
Add to cart
Capacity (English):
0.008 to 0.042 in.
Cut Type:
Semi-flush
Handle Type:
Ergonomic long
Head Style:
Medium Round
Price (MYR)
724.00
EA
Add to cart
  • Induction hardened carbon tool steel
  • Edges hardened to RHC 63-65
  • Dual leaf springs
  • ESD safe cushion grips
Specifications
Handle TypeErgonomic long
GradeUltra Precision
Head StyleMedium Round
Length (English)5.75 in.
Length (Metric)14.6 cm
Material1% Carbon Tool Steel
Capacity (English)0.008 to 0.042 in.
Cut TypeSemi-flush
Grip TypeESD-safe soft foam cushion
IncludesInduction hardened cutting blades to RHC 63-65, Stainless Steel dual leaf springs
Length (English) Head0.44 in.
Length (English) Tip0.43 in.
Length (Metric) Head1.11 cm
Length (Metric) Tip1.09 cm
Unit SizeEA