Check Order Status
Chrome Vanadium Tools
Actual product may vary
Chrome Vanadium Tools
Feature ESD Safe cushion grips
Have Questions?
Catalog Number
Material
ECT51I-6
Chrome Vanadium Steel
Catalog number
ECT51I-6
Price (MYR)
904.00
EA
Add to cart
Material:
Chrome Vanadium Steel
Price (MYR)
904.00
EA
Add to cart
Locking device keeps cutting jaws closed at rest
0.5 in. cutting capacity makes shear cuts
6 in. length
Specifications
Product Type
Cable Cutter
Length (English)
4 in.
Length (Metric)
10.2 cm
Material
Chrome Vanadium Steel
For Use With (Application)
Cutting stranded cables and wire up to 0.5 in. dia.
Includes
Cutting jaws