Check Order Status
Duckbill Point Tweezers
Actual product may vary
Duckbill Point Tweezers
Duckbill Point Tweezers used for handling labels and SMD components.
Have Questions?
Catalog Number
Length (English)
Tip Style
Material
Precision
ECT2A-SA
2.8 in.
Tapered, Flat Square Duckbilled
Anti-magnetic Stainless Steel
High
Catalog number
ECT2A-SA
Price (MYR)
354.00
EA
Add to cart
Length (English):
2.8 in.
Tip Style:
Tapered, Flat Square Duckbilled
Material:
Anti-magnetic Stainless Steel
Precision:
High
Price (MYR)
354.00
EA
Add to cart
Anti-magnetic stainless steel or Carbon Steel or Magnetic Stainless Steel
Specifications
Tip Style
Tapered, Flat Square Duckbilled
Length (English)
2.8 in.
Material
Anti-magnetic Stainless Steel
Length (Metric)
71 mm
Product Type
Tweezers
Quantity (No. per Case)
1 Ea.
For Use With (Application)
For handling labels and SMD components
Height (English) Body
0.015 in.
Height (Metric) Body
0.038 cm
Precision
High
Width (English) Body
0.25 in.
Width (Metric) Body
0.63 cm
Style
High-precision