Check Order Status
Aramid Fiber Scissors
Actual product may vary
Aramid Fiber Scissors
Smooth opening affords easy operation
Have Questions?
Catalog Number
Length (Metric)
Material
ECT299P
139.7 mm
Carbon Steel
Catalog number
ECT299P
Price (MYR)
291.00
EA
Add to cart
Length (Metric):
139.7 mm
Material:
Carbon Steel
Price (MYR)
291.00
EA
Add to cart
Molded handles
Tempered stainless-steel blades
Serrations on one edge
Wider finger holes
Overall length: 5.5 in. (139.7mm)
Blade length 1.75 in. (44.4mm)
Specifications
Blade Style
Straight
Length (English)
5.5 in.
Length (Metric)
139.7 mm
Material
Carbon Steel
For Use With (Application)
Cutting Dupont™ Kevlar™ aramid fiber