Nalgene™ Fluorinated HDPE, Carboy with Spigot
Nalgene™ Fluorinated HDPE, Carboy with Spigot
Actual product may vary
Nalgene™ Fluorinated HDPE, Carboy with Spigot
Nalgene™ Fluorinated HDPE, Carboy with Spigot
Nalgene™ Fluorinated HDPE, Carboy with Spigot
Nalgene™ Fluorinated HDPE, Carboy with Spigot
Nalgene™ Fluorinated HDPE, Carboy with Spigot
For Use With: Ideal for storing and dispensing a variety of solvents; Closure Material: Fluorinated Polypropylene; Color: White
Have Questions?
Catalog NumberCapacity (Metric)
DS2327-005020 L
Catalog number DS2327-0050
Price (MYR)
1,096.00
CS
Add to cart
Capacity (Metric):
20 L
Price (MYR)
1,096.00
CS
Add to cart
Easily fill and clean the Thermo ScientificTM NalgeneTM Rectangular Fluorinated HDPE Carboy with Spigot, which features a wide-mouth opening and a sturdy stainless-steel handle. These Carboys are ideal for storing and dispensing a variety of solvents, and the fluorinated surface (inside and out) improves barrier properties and reduces solvent absorption while enhancing long-term container performance to prevent loss due to permeation.
Features:
  • Molded-in graduations of 1 gallon and 5L increments
  • Molded-in spigot boss provides strength and leak resistance
  • Rectangular shape saves valuable space
  • Graduated
Includes:
ETFE spigot and fluorinated white polypropylene screw closure
Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged.
Specifications
AutoclavableNot Autoclavable
Capacity (English)5.3 gal.
Capacity (Metric)20 L
Closure MaterialFluorinated Polypropylene
Closure Size100 mm
ColorWhite
DescriptionFluorinated Rectangular Carboy w/Spigot FLPE 20 L
Diameter (Metric) Inner Neck86 mm
For Use With (Application)Ideal for storing and dispensing a variety of solvents
Height (Metric) With Closure381 mm
IncludesMolded-in spigot boss
Length (Metric)318 mm
MaterialFluorinated HDPE
No. per Case1 Ea.
No. per Pack1 Ea.
Product TypeCarboy with Spigot
Width (Metric)219 mm
Unit SizeCS
Frequently asked questions (FAQs)
To what level are your FLPE fluorinated containers processed?
All of our fluorinated products are processed to Level 5.