Decomatic
Decomatic
Actual product may vary
Decomatic
Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines
Have Questions?
Catalog NumberVolume (Metric)
DECD002821
also known as DECD/0028/21
5000 mL
Catalog number DECD002821
also known as DECD/0028/21
Price (MYR)
561.00
BOT
Add to cart
Volume (Metric):
5000 mL
Price (MYR)
561.00
BOT
Add to cart
Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines
Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines. By preventing the deposition of lime and salts on both contents and machine parts, it minimises corrosion and wear of seals, pumps, racks and stainless steel surfaces, thereby providing more efficient performance and longer life to the machine. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Ideal for use on glassware and equipment in medical, pharmaceutical, research, educational, scientific and industrial laboratories.
Specifications
FormLiquid
pH>13
AppearanceAmber
Viscosity2.28cps at 20°C
Volume (Metric)5000 mL
Unit SizeBOT