Neutracon
Neutracon
Actual product may vary
Neutracon
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Have Questions?
Catalog NumberVolume (Metric)
DECD002721
also known as DECD/0027/21
5000 mL
Catalog number DECD002721
also known as DECD/0027/21
Price (MYR)
552.00
EA
Add to cart
Volume (Metric):
5000 mL
Price (MYR)
552.00
EA
Add to cart
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Biodegradable, phosphate, enzyme and chlorine bleach free, near-neutral liquid surface-active agent. Completely soluble in water and will not deposit calcium and magnesium or precipitate iron. Solutions are completely stable in air and do not absorb CO2. May be used in conjunction with water soluble solvents. A 13% solution is sufficient for the majority of cleaning applications. Heating the solution, or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Specifically formulated for cleaning/decontamination of non-ferrous metals, 'soft' silica glass, polymer surfaces and other materials which may be corroded, etched, degraded, discolored or weakened by acidic or alkaline cleaning agents.
Specifications
FormLiquid
pH7.5
AppearanceAmber
CompositionPhosphate, Enzyme, Chlorine Bleach Free, Water
Viscosity55.0cps at 23°C
Volume (Metric)5000 mL
Unit SizeEA