Owl™ Casting Dams
Owl™ Casting Dams
Actual product may vary
Owl™ Casting Dams
Owl™ Casting Dams
Owl™ Casting Dams
Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.
Have Questions?
Catalog NumberDepth (Metric)
DAM-2323 cm
Catalog number DAM-23
Price (MYR)
1,299.00
EA
Add to cart
Depth (Metric):
23 cm
Price (MYR)
1,299.00
EA
Add to cart
Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.
  • Seal agarose on contact, eliminating the need for tape
  • Does not attach to gel tray, enabling gels of any length to be poured
  • Easily handled and maneuvered using knob or finger depressions
  • Dam blocks off a portion of UVT gel tray
Specifications
DescriptionCasting Dam, 23cm (L)
For Use With (Equipment)Owl System
Product LineOwl
TypeGel Casting Dam
Depth (Metric)23 cm
Unit SizeEA