Barnstead™ E-Pure™ Optional Accessories
Barnstead™ E-Pure™ Optional Accessories
Actual product may vary
Barnstead™ E-Pure™ Optional Accessories
Barnstead™ E-Pure™ Optional Accessories
Barnstead™ E-Pure™ Optional Accessories
Barnstead™ E-Pure™ Optional Accessories
Barnstead™ E-Pure™ Optional Accessories
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
Have Questions?
Catalog NumberTypeVoltage
D8952Remote Dispenser0/1 V
Catalog number D8952
Price (MYR)
7,451.00
EA
Add to cart
Type:
Remote Dispenser
Voltage:
0/1 V
Price (MYR)
7,451.00
EA
Add to cart
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
Specifications
For Use With (Equipment)Barnstead E-Pure
TypeRemote Dispenser
Voltage0/1 V
Unit SizeEA