Check Order Status
Barnstead™ ROpure LP™ Cartridges
Actual product may vary
Barnstead™ ROpure LP™ Cartridges
Thermo Scientific Barnstead; For Use with: Nanopure Quad, E-pure; Cartridge: Ultrapure Ion Exchange
Have Questions?
Catalog number
D5027
Price (MYR)
1,187.00
EA
Add to cart
Price (MYR)
1,187.00
EA
Add to cart
Thermo Scientific Barnstead; For Use with: Nanopure Quad, E-pure; Cartridge: Ultrapure Ion Exchange
Pretreatment/Prefilter, Thin Film
Extruded carbon cartridge (with 5μm prefilter) extends life of 09-050-37 membrane by removing chlorine from feedwater
Ultrapure Ion Exchange
Capacity: 760 grains as NaCl to 1megohm endpoint
Can also be used with E-pure™ systems and Nanopure™ systems
Specifications
Capacity
760 grains
Purification Method
Ultrapure
For Use With (Application)
Spring ion exchange
For Use With (Equipment)
Nanopure Quad and E-Pure System, Spring Ion Exchange
Type
Cartridge