Barnstead Pretreatment Cartridges
Product Image
Barnstead Pretreatment Cartridges
Barnstead Pretreatment Cartridges
Have Questions?
Catalog number D50250
Price (MYR)
903.00
EA
Add to cart
Price (MYR)
903.00
EA
Add to cart
Barnstead Pretreatment Cartridges
Specifications
Resin TypeActivated Carbon
For Use With (Equipment)Nanopure Infinity System, Replacement Pretreatment Cartridge
TypePretreatment Cartridge
Unit SizeEA