Barnstead™ E-Pure™ Cartridge Kits
Barnstead™ E-Pure™ Cartridge Kits
Actual product may vary
Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
D5023Cartridge kit for E-Pure 4-holder, Tap feed, Organic free
D50227Cartridge kit for E-Pure 4-holder; Pretreated Feed
Catalog number D5023
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Cartridge kit for E-Pure 4-holder, Tap feed, Organic free
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
DescriptionCartridge kit for E-Pure 4-holder, Tap feed, Organic free
Feedwater SourceTap
For Use With (Equipment)Cartridge kit for E-Pure 4-holder, Tap feed, Organic free
TypeCartridge Kit
Unit SizeEA