D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
Actual product may vary
D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs.
Have Questions?
Catalog NumberTeeth CountThickness (Metric) Teeth
D3-MTC25 (1X)1 mm
Catalog number D3-MTC
Price (MYR)
633.00
EA
Add to cart
Teeth Count:
25 (1X)
Thickness (Metric) Teeth:
1 mm
Price (MYR)
633.00
EA
Add to cart
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs. These microwell and wall combs are heavy-duty, one-piece units for use with the D3-14, A3-1 and A6 wide gel electrophoresis systems.
Ordering Information:
  • Cat. Nos. D3-MTC, -MTD, -MT2C and -MT2D are 8 and 12-channel pipette format (1X) and (2X) combs
  • Customized combs are available (Order SKU # XCM); please contact your customer service or sales representative for further details
Specifications
DescriptionMicrowell comb
For Use With (Equipment)Owl™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System
TypeComb
Teeth Count25 (1X)
Thickness (Metric) Teeth1 mm
Unit SizeEA