Owl™ D2 Wide-Gel Electrophoresis System Combs
Owl™ D2 Wide-Gel Electrophoresis System Combs
Actual product may vary
Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTeeth CountThickness (Metric) Teeth
D1-MTD151.5 mm
D1-20D201.5 mm
Catalog number D1-MTD
Price (MYR)
550.00
EA
Add to cart
Teeth Count:
15
Thickness (Metric) Teeth:
1.5 mm
Price (MYR)
550.00
EA
Add to cart
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
For Use With (Equipment)Owl™ D2 Wide Gel Electrophoresis System
Product LineOwl
TypeMicrowell Comb
Teeth Count15
Thickness (Metric) Teeth1.5 mm
Unit SizeEA