Traceable™ Silicone Pump Tubing
Traceable™ Silicone Pump Tubing
Actual product may vary
Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerFor Use With
CTL33661/4 in.Medium flow and Medium/High Flow Variable-Speed Pump
CRG33651/4 in.Ultra Low Flow and Low Flow Variable-Speed Pump
Catalog number CTL3366
Price (MYR)
-
Diameter (English) Inner:
1/4 in.
For Use With:
Medium flow and Medium/High Flow Variable-Speed Pump
Ideal for use with food, pharmaceuticals, and other critical solutions
  • Replacement tubing for Variable-Speed Pump
  • Can be sterilized by autoclaving
Specifications
AutoclavableAutoclavable
Diameter (English) Inner1/4 in.
Diameter (Metric) Inner6.35 mm
MaterialSilicone
Overall Length (English)25 ft.
Overall Length (Metric)7.6 m
For Use WithMedium flow and Medium/High Flow Variable-Speed Pump
Product TypePump Tubing
Unit SizeEA