Check Order Status
Compass Board
Actual product may vary
Compass Board
Great for a simple lines of force demonstration.
Have Questions?
Catalog number
CTL3135
Price (MYR)
-
Ungraduated and transparent
Size: 20mm diameter
Specifications
Color
Transparent
Description
Ungraduated
Weight (English)
Ship Weight: 3 lb.
Product Type
Compass Board