Memory-Loc™ Liquid Nitrogen Traceable™ DataLogger Thermometer
Memory-Loc™ Liquid Nitrogen Traceable™ DataLogger Thermometer
Actual product may vary
Memory-Loc™ Liquid Nitrogen Traceable™ DataLogger Thermometer
Memory-Loc™ Liquid Nitrogen Traceable™ DataLogger Thermometer can be used in 21 CFR 11 environments, raw data is locked and may not be cleared/changed on base unit.
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL15-079-671-196°C to +105°C
Catalog number CTL15-079-671
Price (MYR)
2,086.00
Special offer
Ends: 31-Dec-2025
2,086.00
EA
Add to cart
Temperature Range (Metric):
-196°C to +105°C
Price (MYR)
2,086.00
Special offer
Ends: 31-Dec-2025
2,086.00
EA
Add to cart
Memory-Loc™ Liquid Nitrogen Traceable™ DataLogger Thermometer can be used in 21 CFR 11 environments, raw data is locked and may not be cleared/changed on base unit.
  • Can be used in 21 CFR 11 environments, raw data is locked and may not be cleared/changed on base unit
  • Over 1 million temperature observations
  • Allows user-defined logging interval from every minute to every 24 hours
  • Unable to delete data from memory once stored
  • Indicator light will appear when memory is 95%
  • 4 calibration points -196°C, -80°C, 0°C, 100°C
  • Range: -200⪚C to +105°C
  • Accuracy of ±0.25°C
  • Resolution of 0.01
Specifications
TypeDatalogger
Data Memory525,600 Readings
CalibrationMulti-point Calibration
Sensor Types1 probe
Accuracy (Temperature)±0.25°C
Temperature Resolution0.01°C
Temperature Range (Metric)-196°C to +105°C
Dimensions (L x W x H)2.75 x 4.25 x 0.75 in.
Weight (English)5 oz.
Unit SizeEA