Memory-Loc™ Datalogging Traceable™ Hygrometer
Memory-Loc™ Datalogging Traceable™ Hygrometer
Actual product may vary
Memory-Loc™ Datalogging Traceable™ Hygrometer
Used to read temperature, relative humidity and time. The Memory-Loc™ Datalogging Traceable™ Hygrometer is great for hoods, storerooms, cleanrooms, and other environments that need to be monitored.
Have Questions?
Catalog NumberHumidity Range
CTL15-079-6670 to 95%, non-condensing
Catalog number CTL15-079-667
Price (MYR)
1,440.00
Special offer
Ends: 31-Dec-2025
1,440.00
EA
Add to cart
Humidity Range:
0 to 95%, non-condensing
Price (MYR)
1,440.00
Special offer
Ends: 31-Dec-2025
1,440.00
EA
Add to cart
Used to read temperature, relative humidity and time. The Memory-Loc™ Datalogging Traceable™ Hygrometer is great for hoods, storerooms, cleanrooms, and other environments that need to be monitored.
  • Fixed memory structure, maintains 688,086 humidity and temperature observations
  • Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
  • Stores up to 30 unique alarm events
  • Memory can NOT be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
  • Status indicators - Low battery, memory full, USB data transfer and active alarm state
Specifications
TypeDatalogger
Display TypeDigital
Data Memory2 Years, 2 Minute Interval
CalibrationMulti-point Calibration
Alarm TypesAudible, Visual
Software TypeTraceableLIVE
IncludesProbe, Stand, Velcro™, Magnetic Backing, Wall Mount, Traceable™ Certificate, and Batteries
Certifications/ComplianceTraceable™ Certificate
Accuracy (Relative Humidity)±3% RH between 5 to 75%, otherwise ±5% RH
Accuracy (Temperature)±0.4°C
Humidity Range0 to 95%, non-condensing
Resolution (Relative Humidity)0.1% RH
Temperature Resolution0.1°C
Temperature Range (Metric)0°C to 65°C
Operating Temperature (English)-4°F to +158°F
Operating Temperature (Metric)-20°C to +70°C
Dimensions (L x W x H)5.5 x 3.25 x 1 in.
Height (English)25.4 mm
Height (Metric)1 in.
Length (English)5.5 in.
Length (Metric)139.7 mm
Width (English)3.2 in.
Width (Metric)82.6 mm
Weight (English)6 oz.
Weight (Metric)170.1 g
Unit SizeEA