Excursion-Trac™ Datalogging Traceable™ Hygrometer
Actual product may vary
Excursion-Trac™ Datalogging Traceable™ Hygrometer
Used to read temperature, relative humidity and time. The Excursion-Trac™ Datalogging Traceable™ Hygrometer is great for hoods, storerooms, cleanrooms, and other environments that need to be monitored.
Have Questions?
Catalog NumberHumidity Range
CTL15-079-6650 to 95%
Catalog number CTL15-079-665
Price (MYR)
1,904.00
Each
Humidity Range:
0 to 95%
Price (MYR)
1,904.00
Each
Used to read temperature, relative humidity and time. The Excursion-Trac™ Datalogging Traceable™ Hygrometer is great for hoods, storerooms, cleanrooms, and other environments that need to be monitored.
  • Rolling memory structure, maintains 525,504 humidity and temperature observations
  • Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
  • Stores up to 30 unique alarm events
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data - unit can remain in use while downloading and analyzing data
  • Status indicators - Low battery, memory full, USB data transfer and active alarm state
Specifications
TypeHygrometer Datalogger
Display TypeDigital
Data Memory2 Years, 2 Minute Interval
CalibrationMulti-point Calibration
Alarm TypesAudible, Visual
Software TypeTraceableLIVE
IncludesProbe, stand, Velcro™, magnetic backing, wall mount, Traceable™ Certificate, and batteries
Certifications/ComplianceTraceable™ Certificate
InterfaceUSB
Sensor TypesWired Chip Glass Thermistor
Accuracy (Relative Humidity)±3% RH between 5 to 75%, otherwise ±5% RH
Accuracy (Temperature)±0.4°C
Humidity Range0 to 95%
Resolution (Relative Humidity)0.1% RH
Temperature Resolution0.1°C
Temperature Range (Metric)0°C to 65°C
Operating Temperature (Metric)-20°C to +70°C
Dimensions (L x W x H)5.5 x 3.25 x 1 in.
Weight (English)6 oz.
Unit SizeEach